Order Entry
Export
Orders LinkContactUsLinkComponent
 

UV lamps with filter, E21 series

UV lamp, 500×85×80 mm, 365 nm, 1100 µW/cm², 1×15 W
  CONSVL115-L
 :   CONSORT
UV lamps with filter, E21 series
  CONSVL115-L
 :  CONSORT
UV lamps with filter, E21 series
UV lamps with filter, E21 series

UV lamp, 500×85×80 mm, 365 nm, 1100 µW/cm², 1×15 W

 
990,00 €
   
1 * 1 items
 
 
  • Description:
    UV lamp, 500×85×80 mm
  • Seq:
    02
  • Intensity:
    1100 µW/cm²
  • Output:
    1×15 W
  • Wavelength:
    365 nm
  • Cat. No.:
    CONSVL115-L