UV lamps with filter, E21 series
UV lamp, 200×85×60 mm, 254/365 nm, 400/610 µW/cm², 2×6 W
Catalog # CONSVL6-LC
Supplier: CONSORT
Jump to:
Product Details & Documents
Specifications
- Description:UV lamp, 200×85×60 mm
- Seq:11
- Intensity:400/610 µW/cm²
- Output:2×6 W
- Wavelength:254/365 nm



