Orders LinkContactUsLinkComponent
Counting chambers, Malassez
Counting chambers, Malassez
Cell counting chamber with bright lines, Malassez, 0,2 mm
Catalog # MARI0640630
Supplier:   MARIENFELD
Counting chambers, Malassez
Catalog # MARI0640630
Supplier:  MARIENFELD
Counting chambers, Malassez
Counting chambers, Malassez
Cell counting chamber with bright lines, Malassez, 0,2 mm
Price
73,76 €
Log in for availability
1 * 1 items
Product Details & Documents
Counting chambers specially designed for in vitro diagnostic.
  • Double net ruling
  • Two cover glasses
  • Dark lined without clamps
Specifications
  • Depth of chamber:
    0,2 mm
  • Type:
    Malassez
  • Description:
    Cell counting chamber with bright lines
  • Seq:
    002