VWR®, IOS W-Plan MET Objectives
IOS W-Plan MET objectives (no cover glass)
630-2159
These objectives deliver precise performance without the need of the cover slide, being NCG (no cover glass). They are intended to be used in metallurgical and epi-illumination applications. Ideal for routine laboratory needs.
Designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.
- Magnification:50×/0,75 mm
- Description:IOS W-Plan MET objectives (no cover glass)
- Seq:204