Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1-Komponenten-Kalibriergas SO2 10ppm/N2 58l Aluzylinder 1 * 58 L
Catalog # CALG8A-SO2N01-C4
Supplier:
CALGAZ
Expand image
1-Komponenten-Kalibriergas SO2 10ppm/N2 58l Aluzylinder 1 * 58 L
Catalog # CALG8A-SO2N01-C4
Supplier:
CALGAZ