Overalls, 4500
White, 3XL
Catalog # MMMA4500W3XL
Supplier: 3M
Jump to:
Product Details & Documents
This simple overall made of durable PP fabric is suitable for non hazardous applications where hygiene and cleanliness are important.
- Excellent economic solution to keep the wearer clean or dry
- Breathable and light duty protection
- Two-way zip with storm flap
- Elasticated 2-panel hood, cuffs, ankles and waist
Typical applications include: Domestic DIY use, light duty industrial cleaning and maintenance applications.
Weight: 45 g/m²
Certifications: Cat. I
Specifications
- Clothing Size:3XL
- Colour:White
- Seq:06
- Cat. No.:MMMA4500W3XL
- Model:4500
- Garment application:General Purpose|Laboratory|Industrial
- Fabric:PP
- Weight:45 g/m²



