Orders LinkContactUsLinkComponent
Overalls, 4500
Overalls, 4500
White, XXL
Catalog # MMMA4500W2XL
Supplier:   3M
Overalls, 4500
Catalog # MMMA4500W2XL
Supplier:  3M
Overalls, 4500
Overalls, 4500
White, XXL
Price
117,00 €
Log in for availability
1 * 20 items
Product Details & Documents
This simple overall made of durable PP fabric is suitable for non hazardous applications where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean or dry
  • Breathable and light duty protection
  • Two-way zip with storm flap
  • Elasticated 2-panel hood, cuffs, ankles and waist
Typical applications include: Domestic DIY use, light duty industrial cleaning and maintenance applications.
Weight: 45 g/m²
Certifications: Cat. I
Specifications
  • Clothing Size:
    XXL
  • Colour:
    White
  • Seq:
    05
  • Cat. No.:
    MMMA4500W2XL
  • Model:
    4500
  • Garment application:
    General Purpose|Laboratory|Industrial
  • Fabric:
    PP
  • Weight:
    45 g/m²