ContactUsLinkComponent
qPCR detection of Human Herpesvirus 6 variant A. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HHV6A
Supplier:  PRIMERDESIGN
qPCR detection of Human Herpesvirus 6 variant A. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HHV6A
Supplier:  PRIMERDESIGN