ContactUsLinkComponent
qPCR detection of H Pylori. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous co ntrol, extraction control 1 * 150 Tests
Catalog # PATH-H.PYL
Supplier:  PRIMERDESIGN
qPCR detection of H Pylori. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous co ntrol, extraction control 1 * 150 Tests
Catalog # PATH-H.PYL
Supplier:  PRIMERDESIGN