ContactUsLinkComponent
qPCR detection of Adenovirus type C. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-ADVC
Supplier:  PRIMERDESIGN
qPCR detection of Adenovirus type C. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-ADVC
Supplier:  PRIMERDESIGN