ContactUsLinkComponent
qPCR detection of Fungi Kingdom (including Yeast) and Amoebozoa. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-FUNGI-AMO
Supplier:  PRIMERDESIGN
qPCR detection of Fungi Kingdom (including Yeast) and Amoebozoa. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-FUNGI-AMO
Supplier:  PRIMERDESIGN