qPCR detection of shiga toxin (stx1) producing Escherichia coli. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
qPCR detection of shiga toxin (stx1) producing Escherichia coli. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests