ContactUsLinkComponent
qPCR detection of Babesia divergens. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # PATH-B.DIVERGE
Supplier:  PRIMERDESIGN
qPCR detection of Babesia divergens. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # PATH-B.DIVERGE
Supplier:  PRIMERDESIGN