ContactUsLinkComponent
qPCR detection of all Group 2 Coronavirus genomes. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standa rd curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HCOV-GP2
Supplier:  PRIMERDESIGN
qPCR detection of all Group 2 Coronavirus genomes. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standa rd curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HCOV-GP2
Supplier:  PRIMERDESIGN