Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KROSFLO MODUL 0.2UM PES 0.5MM ID 5.10M2 1 * 1 items
Catalog # SPECK06-P20U-05-N
Supplier:
Spectrum Laboratories
Expand image
KROSFLO MODUL 0.2UM PES 0.5MM ID 5.10M2 1 * 1 items
Catalog # SPECK06-P20U-05-N
Supplier:
Spectrum Laboratories