Orders LinkContactUsLinkComponent
Microplates, 384-well
HiBase, white, medium binding, -
Catalog # GREI784075-25_150
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 384-well
Catalog # GREI784075-25_150
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 384-well
Microplates, 384-well
HiBase, white, medium binding, -
Price
2 010,00 €
Log in for availability
1 * 150 items
Product Details & Documents
PS. With the exception of the Small Volume™, all 384-well PS microplates have rounded square wells, which combine the advantages of flexible working volume (10 to 130 µl) with reduced wicking and bubbling. Small Volume™ plates have round wells with a conical geometry, which allows a working volume of just 4 to 25 µl. All plates are supplied without lid, except for some sterile formats.
  • Flat bottom well profile
  • Choice of solid or µClear® bottom
  • Medium or high binding versions available
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifications
  • Sterile:
    -
  • Description:
    HiBase, white, medium binding
  • Seq:
    17
  • Cat. No.:
    GREI784075-25_150
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact