SPE columns, Supra-Poly® HLB
HLB, 30 µm, 15 ml, 100 mg
Catalog # PERCN9306670
Supplier: PerkinElmer
Jump to:
Product Details & Documents
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
Specifications
- Particle Size:30 µm
- Capacity:15 ml
- Seq:18
- Bed weight:100 mg
- Phase:HLB



