Order Entry
Export
Orders LinkContactUsLinkComponent
SPE columns, Supra-Poly® HLB

SPE columns, Supra-Poly® HLB

HLB, 60 µm, 15 ml, 1 g
Catalog # PERCN9306686
Supplier:   PerkinElmer
SPE columns, Supra-Poly® HLB
Catalog # PERCN9306686
Supplier:  PerkinElmer
SPE columns, Supra-Poly® HLB

HLB, 60 µm, 15 ml, 1 g

Price
864,00 €
Log in for availability
1 * 1 items
Product Details & Documents
  • Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
  • Shorter analysis times, greater load capacity and reduced solvent usage
  • Ideal for high throughput assays
Specifications
  • Particle Size:
    60 µm
  • Capacity:
    15 ml
  • Seq:
    43
  • Bed weight:
    1 g
  • Phase:
    HLB