Order Entry
Export
Orders LinkContactUsLinkComponent
 

Reading magnifiers, powerlux

Reading magnifier, 58 mm
Catalog # 630-2182
Supplier:   ESCHENBACH OPTIK
Reading magnifiers, powerlux
Catalog # 630-2182
Supplier:  ESCHENBACH OPTIK
Reading magnifiers, powerlux
Reading magnifiers, powerlux

Reading magnifier, 58 mm

Price
207,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These illuminated magnifiers have a Ceratec® anti-scratch hard coating ensuring reliability, comfort and a relaxed vision.

  • Comfortable use
  • Automatic switch off after 30 minutes
  • Compact design for intuitive movement on the material

They consistently provide bright and glare-free illumination thanks to two SMD LEDs (8000 K).

Delivery information: Supplied with a protective hard foam case and three AA batteries.

Specifications
  • Ø:
    58 mm
  • Magnification:
  • Description:
    Reading magnifier