Orders LinkContactUsLinkComponent
Labels for BBP3x Printers, B-569
Labels for BBP3x Printers, B-569
High performance label printer tape, B-569, B30C-4000-569-OC, Polyester, Permanent acrylic, Thermal transfer, Ochre, BBP31, 101 mm×30,4 m, 1 roll/box
Catalog # BRDY117957
Supplier:   Brady
Labels for BBP3x Printers, B-569
Catalog # BRDY117957
Supplier:  Brady
Labels for BBP3x Printers, B-569
Labels for BBP3x Printers, B-569
High performance label printer tape, B-569, B30C-4000-569-OC, Polyester, Permanent acrylic, Thermal transfer, Ochre, BBP31, 101 mm×30,4 m, 1 roll/box
Price
456,00 €
Log in for availability
1 * 1 Roll
Product Details & Documents
This high performance polyester labeling tape is used for labeling stainless steel pipes and surfaces. Applies best to very smooth surfaces and ideal for mounting to rigid panels.
Specifications
  • Printer model:
    BBP31
  • Printing method:
    Thermal transfer
  • Type:
    B-569
  • Colour:
    Ochre
  • Description:
    High performance label printer tape
  • Dimensions:
    101 mm×30,4 m
  • Material:
    Polyester
  • Seq:
    45
  • Model:
    B30C-4000-569-OC
  • Packed:
    1 roll/box
  • Adhesive:
    Permanent acrylic