Swabs in tubes
Swabs in tube, White, Plastic/rayon, +
Catalog # GREI425010_1000
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
- Sterilised by irradiation
- Tubes are made of polystyrene
- Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Certificates
Search by Lot NumberSpecifications
- Sterile:+
- Description:Swabs in tube
- Seq:08
- Shaft/head:Plastic/rayon
- Cap colour:White