ContactUsLinkComponent
Dripping-point thermometers, acc. to Ubbelohde, type BE 1/100/230, capillary transparent prismatic, mercury filled, range 100+230C in 1C, immersion length: total, total length: 235 mm, complete with metal ferules DIN 51 801 (3 pieces) Until 10th Oc 1 * 1 items
Catalog # SCNL1262403/200801
undefined
Dripping-point thermometers, acc. to Ubbelohde, type BE 1/100/230, capillary transparent prismatic, mercury filled, range 100+230C in 1C, immersion length: total, total length: 235 mm, complete with metal ferules DIN 51 801 (3 pieces) Until 10th Oc 1 * 1 items
Catalog # SCNL1262403/200801