Order Entry
Export
Orders LinkContactUsLinkComponent
Accessories for Technovit® 9100

Accessories for Technovit® 9100

PMMA powder, 1000 g
Catalog # HKUL66010251
Supplier:   HERAEUS KULZER
Accessories for Technovit® 9100
Catalog # HKUL66010251
Supplier:  HERAEUS KULZER
Accessories for Technovit® 9100

PMMA powder, 1000 g

Price
524,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Documents

Specifications
  • Description:
    PMMA powder, 1000 g
  • Seq:
    902