ContactUsLinkComponent
Epo - Mouse, 4 unique 29mer shRNA constructs in lentiviral GFP vector (Gene ID = 13856). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pGFP-C-shLenti Vector, TR30021, included for free. 1 * 1 KIT
Catalog # ORIGTL500625
Supplier:  OriGene
Epo - Mouse, 4 unique 29mer shRNA constructs in lentiviral GFP vector (Gene ID = 13856). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pGFP-C-shLenti Vector, TR30021, included for free. 1 * 1 KIT
Catalog # ORIGTL500625
Supplier:  OriGene