1,5-Naphthyridine 97+%
250 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 254-79-5 Molecular Formula: C8H6N2 Formula Weight: 130.15 Physical Form: Solid Density: 1.25 Appearance: Off-white Melting Point: 69-75° Beilstein Reference: 109350 MDL No.: MFCD00039727
- CAS Number:254-79-5
- Formula:C₈H₆N₂
- MDL Number:MFCD00039727
- Molecular Weight:130.15 g/mol
- Melting Pt:69…75 °C
- Density:1.25
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pk:250 mg



