Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethylbenzaldehyde 97%

2,3-Dimethylbenzaldehyde 97%

5 g
Catalog # H64425.06
2,3-Dimethylbenzaldehyde 97%
Catalog # H64425.06
CAS Number:  5779-93-1
2,3-Dimethylbenzaldehyde 97%
2,3-Dimethylbenzaldehyde 97%

5 g

CAS Number:  5779-93-1
Price
41,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 5779-93-1 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.029 Refractive Index: 1.5530 Appearance: Colourless to pale yellow Boiling Point: 86-88°/10mm Beilstein Reference: 2238677 MDL No.: MFCD00798004

  • CAS Number:5779-93-1
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00798004
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:86…88 °C (10 mmHg)
  • Density:1.029
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    5 g

Specifications