Orders LinkContactUsLinkComponent
Chicken serum
Chicken serum
Chicken serum, Sterile filtered, Frozen
Catalog # S0500-1000
Supplier:   Biowest
Chicken serum
Catalog # S0500-1000
Supplier:  Biowest
Chicken serum
Chicken serum
Chicken serum, Sterile filtered, Frozen
Price
155,00 €
Log in for availability
1 * 1.000 mL
Product Details & Documents
Chicken serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Chicken serum is used as a blocking agent in immunoassays and controls.
Documents
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Chicken serum
  • Seq:
    17