Orders LinkContactUsLinkComponent
Rabbit plasma
Rabbit plasma with sodium citrate, sterile filtered, 500 ml
Catalog # S2610-500
Rabbit plasma
Catalog # S2610-500
Supplier:  
CAS Number:  
Rabbit plasma
Rabbit plasma with sodium citrate, sterile filtered, 500 ml
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.
Specifications
Specifications
  • Pk:
    500 ml
  • Description:
    Rabbit plasma with sodium citrate, sterile filtered
Specifications