Cellulose chromatography papers
Sheets, 580×600 mm
SARTFT2503-580600N
Chromatography papers are not only ideal for chromatography, but also for routine and repetitive separations, routine analysis of proteins in serum (e.g. human albumin) and several absorption applications like those common in the life sciences and diagnostics. They are made of cotton linters with more than 98% alpha-cellulose content and <0,04% ash content.
- Description:Sheets
- Seq:04
- Size:580×600 mm
- Thickness:0,19 mm
- Cat. No.:SARTFT2503-580600N
- Environmental certification:Renewable Energy Certificate
- Capillary rise:95 mm/30 min
- Grade:FN 3
- Environmentally Preferable:Low Manufacturing Impact
- Weight:90 g/m²



