ContactUsLinkComponent
Horizontal arm standard , with ball bearings for smooth positioning, 598 mm length,with locking ring, grease-free run, adjustable stop for repeatable positioning, adjustable break, interface 15.8 mm (5/8 ) for focussing drives/arms (drives/arms are 1 * 1 items
Catalog # LEIC10447098
Supplier:  LEICA MICROSYSTEMS
Horizontal arm standard , with ball bearings for smooth positioning, 598 mm length,with locking ring, grease-free run, adjustable stop for repeatable positioning, adjustable break, interface 15.8 mm (5/8 ) for focussing drives/arms (drives/arms are 1 * 1 items
Catalog # LEIC10447098
Supplier:  LEICA MICROSYSTEMS