Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, DNA loading buffers

VWR®, DNA loading buffers

Loading Dye Red 5X, 5× 1 ml, 1 KIT
Catalog # 733-2574
Supplier:   VWR Chemicals
VWR®, DNA loading buffers
Catalog # 733-2574
Supplier:  VWR Chemicals
CAS Number:  
VWR®, DNA loading buffers
VWR®, DNA loading buffers

Loading Dye Red 5X, 5× 1 ml, 1 KIT

CAS Number:  
Price
42,70 €
Log in for availability
1 * 1 KIT
Product Details & Documents

These DNA loading dyes are used to load DNA samples to agarose or SDS DNA gels for gel electrophoresis.

Suitable for TAE, TBE, SB and LB electrophoresis buffers.

 : EC Regulation No. 1907/2006 (REACH).

 : Each kit contains 5×1 ml.

 : Store at –20 °C for in vitro use only.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Loading Dye Red 5X
  • Pk Info:
    5× 1 ml

Specifications