Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Mouse SerpinA11 His tag protein. 1 * 100 µG
Catalog # BIRBORB83596-100
Supplier:
Biorbyt
Expand image
Recombinant Mouse SerpinA11 His tag protein. 1 * 100 µG
Catalog # BIRBORB83596-100
Supplier:
Biorbyt
Related Information
Documentation
Datasheet
orb83596
Documentation