Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse recombinant M CSF

Mouse recombinant M CSF

2 µG
Catalog # BIRBORB80519-2
Supplier:   Biorbyt
Mouse recombinant M CSF
Catalog # BIRBORB80519-2
Supplier:  Biorbyt
CAS Number:  
Mouse recombinant M CSF

2 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 2 µG
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    2 µG
  • Protein/peptide name:
    M CSF
  • Protein synonyms:
    Macrophage Colony Stimulating Factor
  • Solubility:
    It is recommended to reconstitute the lyophilized M-CSF in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • UniProtKB:
    P07141
  • Purity:
    >98%
  • Sequence:
    MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
  • Biological activity:
    The ED50 as determined by the dose-dependent stimulation of the proliferation of M-NFS-60 cells is < 1.0 ng/ml, corresponding to a specific activity of > 1 x 106 units/mg.
  • Cat. No.:
    BIRBORB80519-2
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C.
  • Formulation:
    The protein (1mg/ml) was lyophilized with 5mM Tris, pH 8.0 100mM NaCl.
  • Tested applications:
    SDS-PAGE
  • Species:
    Mouse

Specifications