Mouse recombinant M CSF
10 µG
BIRBORB80519-10
: Biorbyt
- Pk:10 µG
- Protein/peptide name:M CSF
- Protein synonyms:Macrophage Colony Stimulating Factor
- Solubility:It is recommended to reconstitute the lyophilized M-CSF in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- UniProtKB:P07141
- Purity:>98%
- Sequence:MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
- Biological activity:The ED50 as determined by the dose-dependent stimulation of the proliferation of M-NFS-60 cells is < 1.0 ng/ml, corresponding to a specific activity of > 1 x 106 units/mg.
- Cat. No.:BIRBORB80519-10
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C.
- Formulation:The protein (1mg/ml) was lyophilized with 5mM Tris, pH 8.0 100mM NaCl.
- Tested applications:SDS-PAGE
- Species:Mouse



