ContactUsLinkComponent
Biotin-conjugated IgG fraction of polyclonal rabbit antiserum to lysozyme from chicken egg white. This product is intended for use i n precipitating and non-precipitating antibody-binding assays (such as e.g., ELISA and Western blotting and 1 * 1 ml
Catalog # BIRBORB20959-1
Supplier:  Biorbyt
Biotin-conjugated IgG fraction of polyclonal rabbit antiserum to lysozyme from chicken egg white. This product is intended for use i n precipitating and non-precipitating antibody-binding assays (such as e.g., ELISA and Western blotting and 1 * 1 ml
Catalog # BIRBORB20959-1
Supplier:  Biorbyt