Anti-LYN Rabbit Polyclonal Antibody
20 µg
Catalog # BIRBORB106346-20
Supplier: Biorbyt
Jump to:
Product Details & Documents
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
orb106346
Datasheet
Specifications
- Size:20 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Concentration:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Molecular weight:53, 56 kDa
- Shipping temperature:Ice
- Antigen name:v-yes-1 Yamaguchi sarcoma viral related oncogene homolog
- Cat. No.:BIRBORB106346-20
- Reactivity:Human|Rat|Mouse
- Immunogen:A synthesized peptide derived from human Lyn
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:LYN
- Storage buffer:Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.



