Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-MYO7A Rabbit Polyclonal Antibody

Anti-MYO7A Rabbit Polyclonal Antibody

200 µg
Catalog # BIRBORB100243-200
Supplier:   Biorbyt
Anti-MYO7A Rabbit Polyclonal Antibody
Catalog # BIRBORB100243-200
Supplier:  Biorbyt
Anti-MYO7A Rabbit Polyclonal Antibody
Anti-MYO7A Rabbit Polyclonal Antibody

200 µg

Price
Inquire For Price
Log in for availability
1 * 200 µG
Product Details & Documents

Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: MYO7A (myosin VIIA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Documents

Specifications
  • Size:
    200 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    purified by affinity chromatography
  • Antibody type:
    Primary
  • Form:
    liquid
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • Concentration:
    0,5 mg/ml
  • Molecular weight:
    220 kDa
  • Shipping temperature:
    Ice
  • Antigen name:
    Myosin 7A
  • Cat. No.:
    BIRBORB100243-200
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    MYO7A
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol