ContactUsLinkComponent
Thermostatically controlled incubator model TC 175 S contains 3 metal racks + 1 bottom grid + 5 sockets. Net capacity appr. 175l. Temp. range 2 to 40C in steps of 0.1C. Ideal for BOD determination at 20 C. Model with standard door.inner size : 470 1 * 1 items
  LOVI2438220
 :  Lovibond
undefined
Thermostatically controlled incubator model TC 175 S contains 3 metal racks + 1 bottom grid + 5 sockets. Net capacity appr. 175l. Temp. range 2 to 40C in steps of 0.1C. Ideal for BOD determination at 20 C. Model with standard door.inner size : 470 1 * 1 items
  LOVI2438220
 :  Lovibond