Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DB-17MS 20M 0.18MM 0.18UM COL TOROID 1 * 1 items
Catalog # HEWL221-4722LTM
Supplier:
Agilent
Expand image
DB-17MS 20M 0.18MM 0.18UM COL TOROID 1 * 1 items
Catalog # HEWL221-4722LTM
Supplier:
Agilent