ContactUsLinkComponent
Micro fluidics 1/16in x .1 union kit. this kit provides leak-free, inert connection from gc valve to cpm system. 1 * 1 items
Catalog # HEWLG1580-61060
Supplier:  Agilent
Micro fluidics 1/16in x .1 union kit. this kit provides leak-free, inert connection from gc valve to cpm system. 1 * 1 items
Catalog # HEWLG1580-61060
Supplier:  Agilent