Order Entry
Export
Orders LinkContactUsLinkComponent
Cleanroom notebooks, BCR® MyNotes

Cleanroom notebooks, BCR® MyNotes

MyNotes Notebooks, Ruled, Memo, 50
Catalog # BERKMNBKMEMOCR
Supplier:   BERKSHIRE
Cleanroom notebooks, BCR® MyNotes
Catalog # BERKMNBKMEMOCR
Supplier:  BERKSHIRE
Cleanroom notebooks, BCR® MyNotes

MyNotes Notebooks, Ruled, Memo, 50

Price
1 120,00 €
Log in for availability
1 * 40 items
Product Details & Documents

BCR® MyNotes Notebooks are an ideal choice for documentation. They are practical for note taking and data recording in the cleanroom (Class 1 - 10,000).

  • Made with Berkshire cleanroom bond 850 (medium 85 g/m² (24 lbs.))
  • Low in particles and fibres
  • Full lot tracking on every case
  • Double packed for cleanliness
  • Smooth surfaces reduce particle contamination
  • Available in college rule (CR) or 1/4" grid options

BCR® Mynotes are made with the purest quality cellulose from specially selected forests and is specially coated to minimise particle and fiber release.

Certifications: Meets RoHS guidelines and designed for use in ISO Class 3-7, EU grade B/C/D.

Specifications
  • Paper size:
    Memo
  • Type:
    Ruled
  • Description:
    MyNotes Notebooks
  • Seq:
    05
  • No. of pages:
    50