Level control systems with signalboxes, SymLine
Signalbox T1 with disc fill level sensor, signal cable 3 meters, hook and loop fastener for disc sensor 2 meters, US-plug
Catalog # SCTV108158
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Signal box T1 with disc fill level sensor, 3 m signal cable, 2 m hook and loop fastener for disc sensor incl. 220/230V Universal power supply unit 1 * 1 SET Catalog #SCTV108125
Product SCTV108125 is a replacement for SCTV108158 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Monitor two filling levels at once, perfect integration into lab furniture with the Built-In Signalbox ant the Signalbox T1 and T4.
- For all types of Safety Waste Caps and other electronic level controllers
- Touchless alarm-resetting fields for two containers
- 2 switch inputs and 2 switch outputs for peripheral units
LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
SCTV108087
Images 360
Specifications
- Description:Signalbox T1 with disc fill level sensor, signal cable 3 meters, hook and loop fastener for disc sensor 2 meters, US-plug
- Seq:07
Accessories
Product Information
- Catalog #AvailabilityPrice
- Signal cable cable for signal transmission, 1,5 mSignal cable cable for signal transmission, 1,5 mSCTV108304Log in for availability1 * 1 items - 31,67 €
Product Information
- Catalog #AvailabilityPrice
- EU power supply
Product Information
- Catalog #AvailabilityPrice
- Dual lock (releasable pressure) closure for disc sensors, 20×20 mmDual lock (releasable pressure) closure for disc sensors, 20×20 mm554-2714Log in for availability1 * 10 items - 30,35 €
Product Information
- Catalog #AvailabilityPrice
- UK power supply
Product Information
- Catalog #AvailabilityPrice
- Velcro strip for disc sensor, 2 m
Product Information
- Catalog #AvailabilityPrice
- Extension cable for signal transmission, 5 m
Product Information
- Catalog #AvailabilityPrice
- Extension cable for signal transmission, 10 mExtension cable for signal transmission, 10 m590-0728Log in for availability1 * 1 items - 50,65 €
Product Information
- Catalog #AvailabilityPrice
- Cable for relay contact, 5 m
Product Information
- Catalog #AvailabilityPrice
- Disc sensor (alarm at empty state)
Product Information
- Catalog #AvailabilityPrice
- Disc sensor (alarm at full state)
Product Information
- Catalog #AvailabilityPrice
- Signal cable for electronic level control
Product Information
- Catalog #AvailabilityPrice
- Holder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HDHolder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HDSCTV106731Log in for availability1 * 1 items - 54,91 €
Product Information
- Catalog #AvailabilityPrice
- Signal lamp for level control, visual signal (flashing) when the level is reached, with magnetic holder for individual attachment, cable length of the lamp 3 m, power supply: 24 V DC EU/UK/US plugSignal lamp for level control, visual signal (flashing) when the level is reached, with magnetic holder for individual attachment, cable length of the lamp 3 m, power supply: 24 V DC EU/UK/US plugSCTV108334Log in for availability1 * 1 items - 218,23 €
Product Information
- Catalog #AvailabilityPrice
- Holder for disc sensor, GL 45
Product Information
- Catalog #AvailabilityPrice
- Signal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectorsSignal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectorsSCTV108334-TLog in for availability- 218,23 €
Product Information
- Catalog #AvailabilityPrice
- Signal lamp (full state) - EU/UK/US, for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment with disc sensor, visual alarm at full state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSignal lamp (full state) - EU/UK/US, for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment with disc sensor, visual alarm at full state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSCTV108335-TLog in for availability1 * 1 items - 404,99 €
Product Information
- Catalog #AvailabilityPrice
- Signal lamp (empty state) - EU/UK/US, for level control, visual and acoustic signal at empty state, with magnetic holder for individual attachment with disc sensor, visual alarm at empty state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSignal lamp (empty state) - EU/UK/US, for level control, visual and acoustic signal at empty state, with magnetic holder for individual attachment with disc sensor, visual alarm at empty state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSCTV108336-TLog in for availability1 * 1 items - 404,99 €



