Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
COLD ZN KIT 1 * 1 KIT
COLD ZN KIT 1 * 1 KIT
Print
COLD ZN KIT 1 * 1 KIT
Catalog # RALA362390-0000
Certificates
Supplier:
RAL
COLD ZN KIT 1 * 1 KIT
Catalog # RALA362390-0000
Supplier:
RAL
CAS Number:
Certificates
Expand image
Return to Top
COLD ZN KIT 1 * 1 KIT
Add to cart
CAS Number:
Price
85,00 €
Log in
for availability
Unit of Measure
1 * 1 KIT
-
+
Add to cart
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Datasheet
RALA362390-0000_TDS_EN