Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
tissue culture plate 24 wells greiner 1 * 100 items
Catalog # GREI662160RIVM
Supplier:
Greiner Bio-One
Expand image
tissue culture plate 24 wells greiner 1 * 100 items
Catalog # GREI662160RIVM
Supplier:
Greiner Bio-One