VWR®, Bouffant Caps, PP
White, 610 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
These caps are made of PP. Suitable for use in non hazardous environments.
- Latex-free
- With clip
Weight: 11 g/m²
Specifications
- Ø:610 mm
- Colour:White
- Cat. No.:113-7919
- Garment application:General Purpose



