Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Bouffant Caps, PP

VWR®, Bouffant Caps, PP

White, 530 mm
Catalog # 113-7918
Supplier:   VWR Collection
VWR®, Bouffant Caps, PP
Catalog # 113-7918
Supplier:  VWR Collection
VWR®, Bouffant Caps, PP
VWR®, Bouffant Caps, PP

White, 530 mm

Price
59,70 €
Log in for availability
1 * 1.000 items
Product Details & Documents

These caps are made of PP. Suitable for use in non hazardous environments.

  • Latex-free
  • With clip

Weight: 11 g/m²

Specifications
  • Ø:
    530 mm
  • Colour:
    White
  • Cat. No.:
    113-7918
  • Garment application:
    General Purpose