Order Entry
Export
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®

1 kg
Catalog # 1914.1000PE
Supplier:   Avantor
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 1914.1000PE
Supplier:  Avantor
CAS Number:  50-81-7
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®

1 kg

CAS Number:  50-81-7
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specification Test Results

  • Assay:
    99.0-100.5 %
  • Appearance of solution:
    passes test
  • Copper (Cu):
    max. 5 ppm
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Identification:
    passes test
  • Iron (Fe):
    max. 2 ppm
  • Oxalic acid:
    passes test
  • Specific Optical Rotation:
    +20.5 - +21.5 °
  • Sulfated Ash :
    max. 0,10 %

Specifications

  • Pk:
    1 kg

Specifications