Order Entry
Export
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER-flex®, TLC Sheets, Precoated

J.T.Baker® BAKER-flex®, TLC Sheets, Precoated

Aluminium oxide (Alox), 200×200 mm, 200 µm, Gypsum, high content
Catalog # 4467-04
Supplier:   Avantor
J.T.Baker® BAKER-flex®, TLC Sheets, Precoated
Catalog # 4467-04
Supplier:  Avantor
J.T.Baker® BAKER-flex®, TLC Sheets, Precoated
J.T.Baker® BAKER-flex®, TLC Sheets, Precoated

Aluminium oxide (Alox), 200×200 mm, 200 µm, Gypsum, high content

Price
614,30 €
Log in for availability
1 * 25 items
Product Details & Documents

Baker-flex® pre-coated flexible polyester sheets have a hard, abrasion resistant coating and solvent resistant substrate for rapid, clean separations. The sheets can be written on and cut to size.

Specifications
  • Coating material:
    Aluminium oxide (Alox)
  • Layer thickness:
    200 µm
  • Plate size:
    200×200 mm
  • Seq:
    06
  • Fluorescent indicator:
    F254
  • Binder:
    Gypsum, high content