Order Entry
Export
Orders LinkContactUsLinkComponent
 

Spray gun

Replacement nozzle
  ENTE500121
 :   ENTEGRIS
Spray gun
  ENTE500121
 :  ENTEGRIS
Spray gun
Spray gun

Replacement nozzle

 
 
   
1 * 1 items
 

High purity PFA and PTFE (all wetted parts).

  • Orifice: 4,8 mm
  • Connection: 1/4" or 1/2" FNPT
  • Pressure: max. 5516 mbar

Used in fume hoods and wet processing stations to help protect against corrosion.

 
  • Description:
    Replacement nozzle
  • Seq:
    08