Nitrogen spray gun
Nitrogen spray gun, blue
Catalog # ENTE421-42-21
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Long lasting and reliable PFA nitrogen spray guns with 0,5 µm PTFE membrane for use in high purity and corrosive chemical applications.
- Integral filter enables air to be filtered before nitrogen is dispensed
- Vented cap/nozzle meets OSHA safety specifications
- PFA construction enables use in high purity and corrosive applications
- Colored caps enable visual chemical differentiation at a glance
Nitrogen cap/nozzle prevents pressure build-up if outlet is blocked.
Orifice 4,8 mm
Max. pressure 75 psig (517 kPa)
Connection ¹/₄" FNPT
Accessories information: Repair kits are available.
Specifications
- Description:Nitrogen spray gun, blue
Accessories
Product Information
- Catalog #AvailabilityPrice
- Repair tool to access front and rear diaphragm of gunRepair tool to access front and rear diaphragm of gunENTE213-80Log in for availability1 * 1 items
Product Information
- Catalog #AvailabilityPrice
- Replacement cap/nozzle, blue
Product Information
- Catalog #AvailabilityPrice
- Replacement cap/nozzle, natural
Product Information
- Catalog #AvailabilityPrice
- Replacement cap/nozzle, orange



